Assay Details
Gene Symbol: nxf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275024.1 | NP_001261953.1 | 2 - 3 | 801 | 57 | |
NM_079387.6 | NP_524111.3 | 3 - 4 | 893 | 57 | ||
NM_168674.4 | NP_730179.2 | 2 - 3 | 801 | 57 | ||
GenBank mRNA | AJ312282.1 | - | 2 - 3 | 704 | 57 | |
AY075582.1 | - | 3 - 4 | 874 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
