Assay Details
Gene Symbol: nxf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275024.1 | NP_001261953.1 | 4 - 5 | 1083 | 82 | |
NM_079387.6 | NP_524111.3 | 5 - 6 | 1175 | 82 | ||
NM_168674.4 | NP_730179.2 | 4 - 5 | 1083 | 82 | ||
GenBank mRNA | AJ312282.1 | - | 4 - 5 | 986 | 82 | |
AY075582.1 | - | 5 - 6 | 1156 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
