Assay Details
Gene Symbol: CycT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275063.2 | NP_001261992.1 | 2 - 3 | 1344 | 68 | |
NM_079403.4 | NP_524127.2 | 2 - 3 | 1344 | 68 | ||
GenBank mRNA | BT150266.1 | - | 2 - 3 | 1282 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
