Assay Details
| Gene Symbol: mus304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079411.4 | NP_524135.3 | 2 - 3 | 2238 | 80 | |
| GenBank mRNA | AF224715.1 | - | 2 - 3 | 2216 | 80 | |
| AY118585.1 | - | 2 - 3 | 2216 | 80 | ||
More Information
Set Membership: |
|

mus304
Genomic Map