Assay Details
Gene Symbol: Kap-alpha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079443.2 | NP_524167.1 | 4 - 5 | 1065 | 90 | |
GenBank mRNA | AF074957.1 | - | 3 - 4 | 953 | 90 | |
BT083428.1 | - | 4 - 5 | 1053 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
