Assay Details
| Gene Symbol: gig | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275160.1 | NP_001262089.1 | 14 - 15 | 5539 | 82 | |
| NM_079453.3 | NP_524177.1 | 14 - 15 | 5548 | 82 | ||
More Information
Set Membership: |
|

gig
Genomic Map