Assay Details
| Gene Symbol: mRpL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079461.3 | NP_524185.1 | 1 - 2 | 227 | 67 | |
| GenBank mRNA | BT050572.1 | - | 1 - 2 | 130 | 67 | |
More Information
Set Membership: |
|

mRpL15
Genomic Map