Assay Details
Gene Symbol: Ac78C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259928.2 | NP_001246857.1 | 6 - 7 | 866 | 57 | |
NM_001275208.1 | NP_001262137.1 | 12 - 13 | 2665 | 57 | ||
NM_001275209.1 | NP_001262138.1 | 13 - 14 | 2876 | 57 | ||
NM_001275210.1 | NP_001262139.1 | 13 - 14 | 2873 | 57 | ||
NM_079470.5 | NP_524194.3 | 13 - 14 | 2876 | 57 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
