Assay Details
Gene Symbol: Ac78C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259928.2 | NP_001246857.1 | 11 - 12 | 2579 | 92 | |
NM_001275208.1 | NP_001262137.1 | 17 - 18 | 4378 | 92 | ||
NM_001275209.1 | NP_001262138.1 | 18 - 19 | 4589 | 92 | ||
NM_001275210.1 | NP_001262139.1 | 18 - 19 | 4586 | 92 | ||
NM_079470.5 | NP_524194.3 | 18 - 19 | 4589 | 92 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
