Assay Details
Gene Symbol: Ac78C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259928.2 | NP_001246857.1 | 12 - 13 | 2677 | 67 | |
NM_001275208.1 | NP_001262137.1 | 18 - 19 | 4476 | 67 | ||
NM_001275209.1 | NP_001262138.1 | 19 - 20 | 4687 | 67 | ||
NM_001275210.1 | NP_001262139.1 | 19 - 20 | 4684 | 67 | ||
NM_079470.5 | NP_524194.3 | 19 - 20 | 4687 | 67 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
