Assay Details
| Gene Symbol: Ac78C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259928.2 | NP_001246857.1 | 16 - 17 | 3630 | 77 | |
| NM_001275208.1 | NP_001262137.1 | 22 - 23 | 5429 | 77 | ||
| NM_001275209.1 | NP_001262138.1 | 23 - 24 | 5640 | 77 | ||
| NM_001275210.1 | NP_001262139.1 | 23 - 24 | 5637 | 77 | ||
| NM_079470.5 | NP_524194.3 | 23 - 24 | 5640 | 77 | ||
More Information
Set Membership: |
|

Ac78C
Genomic Map