Assay Details
Gene Symbol: Ac78C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275208.1 | NP_001262137.1 | 3 - 4 | 1169 | 67 | |
NM_001275209.1 | NP_001262138.1 | 4 - 5 | 1380 | 67 | ||
NM_001275210.1 | NP_001262139.1 | 4 - 5 | 1377 | 67 | ||
NM_079470.5 | NP_524194.3 | 4 - 5 | 1380 | 67 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
