Assay Details
Gene Symbol: CycH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079483.3 | NP_524207.1 | 4 - 5 | 869 | 77 | |
GenBank mRNA | AF074334.1 | - | 5 - 6 | 940 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
