Assay Details
| Gene Symbol: Chs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144525.2 | NP_001137997.2 | 4 - 5 | 616 | 108 | |
| NM_079485.4 | NP_524209.3 | 4 - 5 | 619 | 108 | ||
More Information
Set Membership: |
|

Chs2
Genomic Map