Assay Details
| Gene Symbol: Hem | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079490.4 | NP_524214.1 | 1 - 2 | 1387 | 70 | |
| GenBank mRNA | AY118579.1 | - | 1 - 2 | 1374 | 70 | |
| X80028.1 | - | 1384 | 70 | |||
More Information
Set Membership: |
|

Hem
Genomic Map