Assay Details
Gene Symbol: alpha-Cat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300225.1 | NP_001287154.1 | 9 - 10 | 3011 | 153 | |
NM_079495.3 | NP_524219.1 | 10 - 11 | 2524 | 153 | ||
GenBank mRNA | D13964.2 | - | 10 - 11 | 2487 | 153 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
