Assay Details
| Gene Symbol: Mos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136973.2 | NP_610817.1 | 2 - 3 | 373 | 61 | |
| GenBank mRNA | BT025923.1 | - | 2 - 3 | 362 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mos| Gene Symbol: Mos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136973.2 | NP_610817.1 | 2 - 3 | 373 | 61 | |
| GenBank mRNA | BT025923.1 | - | 2 - 3 | 362 | 61 | |
Set Membership: |
|