Assay Details
Gene Symbol: mRpL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136974.3 | NP_610818.1 | 1 | 674 | 74 | |
GenBank mRNA | AY118885.1 | - | 1 | 709 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
