Assay Details
Gene Symbol: Sans | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136985.3 | NP_610829.1 | 4 - 5 | 641 | 71 | |
NM_176160.2 | NP_788340.1 | 4 - 5 | 634 | 71 | ||
GenBank mRNA | BT009925.1 | - | 4 - 5 | 597 | 71 |

More Information
Set Membership: |
![]() ![]() |
