Assay Details
| Gene Symbol: Sans | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136985.3 | NP_610829.1 | 5 - 6 | 1645 | 67 | |
| NM_176160.2 | NP_788340.1 | 5 - 6 | 1638 | 67 | ||
| GenBank mRNA | BT009925.1 | - | 5 - 6 | 1601 | 67 | |
More Information
Set Membership: |
|

Sans
Genomic Map