Assay Details
Gene Symbol: cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259368.2 | NP_001246297.1 | 1 - 2 | 416 | 66 | |
NM_001259369.2 | NP_001246298.1 | 1 - 2 | 416 | 66 | ||
NM_137031.3 | NP_610875.1 | 1 - 2 | 416 | 66 | ||
GenBank mRNA | AY051853.1 | - | 1 - 2 | 338 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
