Assay Details
| Gene Symbol: cbc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137032.4 | NP_610876.1 | 1 - 2 | 201 | 68 | |
| GenBank mRNA | AY061187.1 | - | 1 - 2 | 206 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cbc| Gene Symbol: cbc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137032.4 | NP_610876.1 | 1 - 2 | 201 | 68 | |
| GenBank mRNA | AY061187.1 | - | 1 - 2 | 206 | 68 | |
Set Membership: |
|