Assay Details
| Gene Symbol: cbc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137032.4 | NP_610876.1 | 3 - 4 | 1262 | 79 | |
| GenBank mRNA | AY061187.1 | - | 3 - 4 | 1267 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cbc| Gene Symbol: cbc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137032.4 | NP_610876.1 | 3 - 4 | 1262 | 79 | |
| GenBank mRNA | AY061187.1 | - | 3 - 4 | 1267 | 79 | |
Set Membership: |
|