Assay Details
Gene Symbol: Lap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202009.2 | NP_001188938.1 | 4 - 5 | 997 | 77 | |
NM_137163.5 | NP_611007.1 | 4 - 5 | 980 | 77 | ||
GenBank mRNA | BT011365.1 | - | 4 - 5 | 981 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
