Assay Details
| Gene Symbol: GPHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137172.4 | NP_611016.2 | 5 - 6 | 1074 | 127 | |
| GenBank mRNA | BT023838.1 | - | 5 - 6 | 1050 | 127 | |
More Information
Set Membership: |
|

GPHR
Genomic Map