Assay Details
Gene Symbol: Zasp52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137214.5 | NP_611058.4 | 12 - 13 | 3517 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
