Assay Details
Gene Symbol: Zasp52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032249.4 | NP_001027420.2 | 13 - 14 | 6466 | 73 | |
NM_001032250.3 | NP_001027421.1 | 13 - 14 | 2224 | 73 | ||
NM_001043086.3 | NP_001036551.1 | 14 - 15 | 2554 | 73 | ||
NM_001274076.1 | NP_001261005.1 | 14 - 15 | 2575 | 73 | ||
NM_001274077.1 | NP_001261006.1 | 12 - 13 | 3838 | 73 | ||
NM_001274078.1 | NP_001261007.1 | 13 - 14 | 3685 | 73 | ||
NM_001274081.1 | NP_001261010.1 | 5 - 6 | 5375 | 73 | ||
NM_001274082.1 | NP_001261011.1 | 13 - 14 | 2050 | 73 | ||
NM_001274083.1 | NP_001261012.1 | 13 - 14 | 2245 | 73 | ||
NM_001274084.1 | NP_001261013.1 | 12 - 13 | 1837 | 73 | ||
NM_001274085.1 | NP_001261014.1 | 12 - 13 | 2032 | 73 | ||
NM_001274086.1 | NP_001261015.1 | 14 - 15 | 2749 | 73 | ||
NM_001299538.1 | NP_001286467.1 | 12 - 13 | 6136 | 73 | ||
NM_001316421.1 | NP_001303350.1 | 12 - 13 | 6310 | 73 | ||
NM_001316422.1 | NP_001303351.1 | 6 - 7 | 1289 | 73 | ||
NM_137214.5 | NP_611058.4 | 13 - 14 | 3697 | 73 | ||
NM_145757.3 | NP_665700.2 | 14 - 15 | 3186 | 73 | ||
GenBank mRNA | BT001752.1 | - | 13 - 14 | 2221 | 73 | |
BT022171.2 | - | 14 - 15 | 2534 | 73 | ||
JN034424.1 | - | 13 - 14 | 2046 | 73 | ||
JN034425.1 | - | 13 - 14 | 2225 | 73 | ||
JN034426.1 | - | 12 - 13 | 1824 | 73 | ||
JN034427.1 | - | 12 - 13 | 2028 | 73 | ||
JN034429.1 | - | 14 - 15 | 2736 | 73 | ||
JN034430.1 | - | 5 - 6 | 5295 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
