Assay Details
Gene Symbol: Zasp52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032249.4 | NP_001027420.2 | 14 - 15 | 6709 | 61 | |
NM_001032250.3 | NP_001027421.1 | 14 - 15 | 2467 | 61 | ||
NM_001043086.3 | NP_001036551.1 | 15 - 16 | 2797 | 61 | ||
NM_001274076.1 | NP_001261005.1 | 15 - 16 | 2818 | 61 | ||
NM_001274077.1 | NP_001261006.1 | 13 - 14 | 4081 | 61 | ||
NM_001274078.1 | NP_001261007.1 | 14 - 15 | 3928 | 61 | ||
NM_001274081.1 | NP_001261010.1 | 6 - 7 | 5618 | 61 | ||
NM_001274082.1 | NP_001261011.1 | 14 - 15 | 2293 | 61 | ||
NM_001274083.1 | NP_001261012.1 | 14 - 15 | 2488 | 61 | ||
NM_001274084.1 | NP_001261013.1 | 13 - 14 | 2080 | 61 | ||
NM_001274085.1 | NP_001261014.1 | 13 - 14 | 2275 | 61 | ||
NM_001274086.1 | NP_001261015.1 | 15 - 16 | 2992 | 61 | ||
NM_001299538.1 | NP_001286467.1 | 13 - 14 | 6379 | 61 | ||
NM_001316421.1 | NP_001303350.1 | 13 - 14 | 6553 | 61 | ||
NM_001316422.1 | NP_001303351.1 | 7 - 8 | 1532 | 61 | ||
NM_137214.5 | NP_611058.4 | 14 - 15 | 3940 | 61 | ||
NM_145757.3 | NP_665700.2 | 15 - 16 | 3429 | 61 | ||
GenBank mRNA | BT001752.1 | - | 14 - 15 | 2464 | 61 | |
BT022171.2 | - | 15 - 16 | 2777 | 61 | ||
JN034424.1 | - | 14 - 15 | 2289 | 61 | ||
JN034425.1 | - | 14 - 15 | 2468 | 61 | ||
JN034426.1 | - | 13 - 14 | 2067 | 61 | ||
JN034427.1 | - | 13 - 14 | 2271 | 61 | ||
JN034429.1 | - | 15 - 16 | 2979 | 61 | ||
JN034430.1 | - | 6 - 7 | 5538 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
