Assay Details
Gene Symbol: Zasp52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032249.4 | NP_001027420.2 | 6 - 7 | 856 | 83 | |
NM_001169692.2 | NP_001163163.1 | 6 - 7 | 856 | 83 | ||
NM_001274076.1 | NP_001261005.1 | 6 - 7 | 856 | 83 | ||
NM_001274077.1 | NP_001261006.1 | 6 - 7 | 856 | 83 | ||
NM_001274082.1 | NP_001261011.1 | 6 - 7 | 856 | 83 | ||
NM_001274083.1 | NP_001261012.1 | 6 - 7 | 856 | 83 | ||
NM_001274084.1 | NP_001261013.1 | 6 - 7 | 856 | 83 | ||
NM_001274085.1 | NP_001261014.1 | 6 - 7 | 856 | 83 | ||
NM_001299538.1 | NP_001286467.1 | 6 - 7 | 856 | 83 | ||
NM_001316423.1 | NP_001303352.1 | 6 - 7 | 912 | 83 | ||
NM_137214.5 | NP_611058.4 | 6 - 7 | 856 | 83 | ||
NM_145757.3 | NP_665700.2 | 6 - 7 | 912 | 83 | ||
GenBank mRNA | BT081376.1 | - | 6 - 7 | 834 | 83 | |
JN034423.1 | - | 6 - 7 | 717 | 83 | ||
JN034424.1 | - | 6 - 7 | 852 | 83 | ||
JN034425.1 | - | 6 - 7 | 836 | 83 | ||
JN034426.1 | - | 6 - 7 | 843 | 83 | ||
JN034427.1 | - | 6 - 7 | 852 | 83 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
