Assay Details
Gene Symbol: Zasp52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032249.4 | NP_001027420.2 | 8 - 9 | 1266 | 59 | |
NM_001032250.3 | NP_001027421.1 | 8 - 9 | 1440 | 59 | ||
NM_001043086.3 | NP_001036551.1 | 8 - 9 | 1440 | 59 | ||
NM_001169692.2 | NP_001163163.1 | 8 - 9 | 1266 | 59 | ||
NM_001274076.1 | NP_001261005.1 | 8 - 9 | 1266 | 59 | ||
NM_001274077.1 | NP_001261006.1 | 8 - 9 | 1266 | 59 | ||
NM_001274078.1 | NP_001261007.1 | 8 - 9 | 1440 | 59 | ||
NM_001274079.1 | NP_001261008.1 | 8 - 9 | 1440 | 59 | ||
NM_001274082.1 | NP_001261011.1 | 8 - 9 | 1266 | 59 | ||
NM_001274083.1 | NP_001261012.1 | 8 - 9 | 1266 | 59 | ||
NM_001274084.1 | NP_001261013.1 | 8 - 9 | 1266 | 59 | ||
NM_001274085.1 | NP_001261014.1 | 8 - 9 | 1266 | 59 | ||
NM_001274086.1 | NP_001261015.1 | 8 - 9 | 1440 | 59 | ||
NM_001299538.1 | NP_001286467.1 | 8 - 9 | 1266 | 59 | ||
NM_001316421.1 | NP_001303350.1 | 8 - 9 | 1440 | 59 | ||
NM_001316423.1 | NP_001303352.1 | 8 - 9 | 1322 | 59 | ||
NM_137214.5 | NP_611058.4 | 8 - 9 | 1266 | 59 | ||
NM_145757.3 | NP_665700.2 | 8 - 9 | 1322 | 59 | ||
GenBank mRNA | BT001752.1 | - | 8 - 9 | 1437 | 59 | |
BT022171.2 | - | 8 - 9 | 1420 | 59 | ||
BT081376.1 | - | 8 - 9 | 1244 | 59 | ||
JN034423.1 | - | 8 - 9 | 1127 | 59 | ||
JN034424.1 | - | 8 - 9 | 1262 | 59 | ||
JN034425.1 | - | 8 - 9 | 1246 | 59 | ||
JN034426.1 | - | 8 - 9 | 1253 | 59 | ||
JN034427.1 | - | 8 - 9 | 1262 | 59 | ||
JN034428.1 | - | 8 - 9 | 1408 | 59 | ||
JN034429.1 | - | 8 - 9 | 1427 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
