Assay Details
Gene Symbol: Zasp52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032249.4 | NP_001027420.2 | 12 - 13 | 6285 | 127 | |
NM_001032250.3 | NP_001027421.1 | 12 - 13 | 2043 | 127 | ||
NM_001043086.3 | NP_001036551.1 | 13 - 14 | 2373 | 127 | ||
NM_001274076.1 | NP_001261005.1 | 13 - 14 | 2394 | 127 | ||
NM_001274077.1 | NP_001261006.1 | 11 - 12 | 3657 | 127 | ||
NM_001274078.1 | NP_001261007.1 | 12 - 13 | 3504 | 127 | ||
NM_001274081.1 | NP_001261010.1 | 4 - 5 | 5194 | 127 | ||
NM_001274082.1 | NP_001261011.1 | 12 - 13 | 1869 | 127 | ||
NM_001274083.1 | NP_001261012.1 | 12 - 13 | 2064 | 127 | ||
NM_001274084.1 | NP_001261013.1 | 11 - 12 | 1656 | 127 | ||
NM_001274085.1 | NP_001261014.1 | 11 - 12 | 1851 | 127 | ||
NM_001274086.1 | NP_001261015.1 | 13 - 14 | 2568 | 127 | ||
NM_001299538.1 | NP_001286467.1 | 11 - 12 | 5955 | 127 | ||
NM_001316421.1 | NP_001303350.1 | 11 - 12 | 6129 | 127 | ||
NM_001316422.1 | NP_001303351.1 | 5 - 6 | 1108 | 127 | ||
NM_145757.3 | NP_665700.2 | 13 - 14 | 3005 | 127 | ||
GenBank mRNA | BT001752.1 | - | 12 - 13 | 2040 | 127 | |
BT022171.2 | - | 13 - 14 | 2353 | 127 | ||
JN034424.1 | - | 12 - 13 | 1865 | 127 | ||
JN034425.1 | - | 12 - 13 | 2044 | 127 | ||
JN034426.1 | - | 11 - 12 | 1643 | 127 | ||
JN034427.1 | - | 11 - 12 | 1847 | 127 | ||
JN034429.1 | - | 13 - 14 | 2555 | 127 | ||
JN034430.1 | - | 4 - 5 | 5114 | 127 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
