Assay Details
| Gene Symbol: Fas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272306.1 | NP_001259235.1 | 5 - 6 | 957 | 91 | |
| NM_001272308.2 | NP_001259237.1 | 6 - 7 | 1002 | 91 | ||
| NM_001297925.1 | NP_001284854.1 | 5 - 6 | 957 | 91 | ||
| NM_001297926.1 | NP_001284855.1 | 5 - 6 | 957 | 91 | ||
| NM_080327.3 | NP_525066.1 | 5 - 6 | 957 | 91 | ||
| NM_167004.2 | NP_726911.1 | 5 - 6 | 957 | 91 | ||
| NM_167005.2 | NP_726912.1 | 5 - 6 | 957 | 91 | ||
| GenBank mRNA | BT014661.1 | - | 5 - 6 | 940 | 91 | |
| M77165.1 | - | 5 - 6 | 898 | 91 | ||
| M77166.1 | - | 5 - 6 | 898 | 91 | ||
More Information
Set Membership: |
|

Fas2
Genomic Map