Assay Details
| Gene Symbol: Fas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167004.2 | NP_726911.1 | 7 - 8 | 2630 | 104 | |
| GenBank mRNA | BT014661.1 | - | 7 - 8 | 2613 | 104 | |
More Information
Set Membership: |
|

Fas2
Genomic Map