Assay Details
| Gene Symbol: Cbp80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080011.3 | NP_524750.2 | 6 - 7 | 2565 | 81 | |
| NM_167012.3 | NP_726938.1 | 6 - 7 | 2600 | 81 | ||
| GenBank mRNA | AY051796.1 | - | 6 - 7 | 2540 | 81 | |
More Information
Set Membership: |
|

Cbp80
Genomic Map