Assay Details
Gene Symbol: fzr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080113.3 | NP_524852.2 | 2 - 3 | 593 | 60 | |
NM_167013.2 | NP_726941.1 | 2 - 3 | 499 | 60 | ||
GenBank mRNA | BT001403.1 | - | 2 - 3 | 1185 | 60 | |
BT010011.1 | - | 2 - 3 | 643 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
