Assay Details
Gene Symbol: rg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042796.3 | NP_001036261.3 | 15 - 16 | 3271 | 67 | |
NM_001144685.2 | NP_001138157.2 | 15 - 16 | 3583 | 67 | ||
NM_001144686.2 | NP_001138158.2 | 15 - 16 | 3613 | 67 | ||
NM_001144687.2 | NP_001138159.2 | 14 - 15 | 2992 | 67 | ||
NM_001297949.1 | NP_001284878.1 | 18 - 19 | 7600 | 67 | ||
NM_001297950.1 | NP_001284879.1 | 15 - 16 | 3613 | 67 | ||
NM_167028.3 | NP_726978.2 | 15 - 16 | 4323 | 67 |
Gene Symbol: CG15465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297948.1 | NP_001284877.1 | 18 - 19 | 7600 | 67 | |
NM_131978.3 | NP_572206.1 | 15 - 16 | 4323 | 67 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
