Assay Details
| Gene Symbol: rg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042796.3 | NP_001036261.3 | 22 - 23 | 7110 | 85 | |
| NM_001297950.1 | NP_001284879.1 | 22 - 23 | 7452 | 85 | ||
More Information
Set Membership: |
|

rg
Genomic Map