Assay Details
Gene Symbol: rg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042796.3 | NP_001036261.3 | 28 - 29 | 11691 | 53 | |
NM_001144685.2 | NP_001138157.2 | 27 - 28 | 11046 | 53 | ||
NM_001144686.2 | NP_001138158.2 | 28 - 29 | 11244 | 53 | ||
NM_001144687.2 | NP_001138159.2 | 26 - 27 | 10455 | 53 | ||
NM_001297949.1 | NP_001284878.1 | 30 - 31 | 15063 | 53 | ||
NM_001297950.1 | NP_001284879.1 | 29 - 30 | 12201 | 53 | ||
NM_167028.3 | NP_726978.2 | 27 - 28 | 11786 | 53 |
Gene Symbol: CG15465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297948.1 | NP_001284877.1 | 30 - 31 | 15063 | 53 | |
NM_131978.3 | NP_572206.1 | 27 - 28 | 11786 | 53 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
