Assay Details
| Gene Symbol: rg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042796.3 | NP_001036261.3 | 10 - 11 | 2471 | 69 | |
| NM_001144685.2 | NP_001138157.2 | 10 - 11 | 2783 | 69 | ||
| NM_001144686.2 | NP_001138158.2 | 10 - 11 | 2783 | 69 | ||
| NM_001297949.1 | NP_001284878.1 | 13 - 14 | 6800 | 69 | ||
| NM_001297950.1 | NP_001284879.1 | 10 - 11 | 2783 | 69 | ||
| Gene Symbol: CG15465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297948.1 | NP_001284877.1 | 13 - 14 | 6800 | 69 | |
More Information
Set Membership: |
|

rg
Genomic Map