Assay Details
Gene Symbol: APC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132101.4 | NP_572329.3 | 3 - 4 | 1283 | 79 | |
NM_167079.2 | NP_727101.1 | 2 - 3 | 1258 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
