Assay Details
| Gene Symbol: Fum1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132111.3 | NP_572339.1 | 4 - 5 | 1332 | 59 | |
| NM_167081.3 | NP_727108.1 | 4 - 5 | 1260 | 59 | ||
| GenBank mRNA | BT099624.1 | - | 4 - 5 | 1224 | 59 | |
| BT126329.1 | - | 4 - 5 | 1299 | 59 | ||
More Information
Set Membership: |
|

Fum1
Genomic Map