Assay Details
Gene Symbol: Pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031878.2 | NP_001027049.1 | 5 - 6 | 2588 | 72 | |
NM_001031879.3 | NP_001027050.1 | 4 - 5 | 2615 | 72 | ||
NM_001031880.3 | NP_001027051.1 | 5 - 6 | 2564 | 72 | ||
NM_001031881.3 | NP_001027052.1 | 5 - 6 | 2558 | 72 | ||
NM_001031882.3 | NP_001027053.1 | 5 - 6 | 2467 | 72 | ||
NM_001031883.3 | NP_001027054.1 | 5 - 6 | 2559 | 72 | ||
NM_001298023.1 | NP_001284952.1 | 4 - 5 | 2101 | 72 | ||
NM_132112.5 | NP_572340.2 | 5 - 6 | 2553 | 72 | ||
NM_167083.4 | NP_727110.1 | 5 - 6 | 2473 | 72 | ||
GenBank mRNA | BT133188.1 | - | 5 - 6 | 2551 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
