Assay Details
| Gene Symbol: dec-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258641.1 | NP_001245570.1 | 4 - 5 | 2978 | 66 | |
| NM_167132.2 | NP_727202.1 | 4 - 5 | 3029 | 66 | ||
| NM_167133.2 | NP_727203.1 | 4 - 5 | 3043 | 66 | ||
More Information
Set Membership: |
|

dec-1
Genomic Map