Assay Details
Gene Symbol: Imp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169246.2 | NP_001162717.1 | 2 - 3 | 1241 | 79 | |
NM_167249.3 | NP_727451.1 | 2 - 3 | 880 | 79 | ||
NM_167250.3 | NP_727452.1 | 3 - 4 | 1031 | 79 | ||
NM_167251.3 | NP_727453.1 | 2 - 3 | 1241 | 79 | ||
NM_167252.3 | NP_727454.1 | 2 - 3 | 1345 | 79 | ||
NM_167253.3 | NP_727455.1 | 3 - 4 | 1237 | 79 | ||
GenBank mRNA | BT133404.1 | - | 2 - 3 | 1238 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
