Assay Details
Gene Symbol: Klp10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298180.1 | NP_001285109.1 | 4 - 5 | 2530 | 55 | |
NM_132459.4 | NP_572687.1 | 4 - 5 | 2442 | 55 | ||
NM_167267.2 | NP_727491.1 | 4 - 5 | 2817 | 55 | ||
NM_167268.2 | NP_727492.1 | 3 - 4 | 2606 | 55 | ||
NM_167269.2 | NP_727493.1 | 4 - 5 | 2438 | 55 | ||
NM_167270.2 | NP_727494.1 | 4 - 5 | 2442 | 55 | ||
GenBank mRNA | AY051763.1 | - | 4 - 5 | 2435 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
