Assay Details
Gene Symbol: Tango10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272532.2 | NP_001259461.1 | 2 - 3 | 731 | 64 | |
NM_132515.3 | NP_572743.1 | 1 - 2 | 543 | 64 | ||
NM_167302.3 | NP_727555.1 | 1 - 2 | 782 | 64 | ||
GenBank mRNA | AY069304.1 | - | 1 - 2 | 539 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
