Assay Details
Gene Symbol: Tango10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272532.2 | NP_001259461.1 | 3 - 4 | 1718 | 72 | |
NM_132515.3 | NP_572743.1 | 2 - 3 | 1530 | 72 | ||
NM_167302.3 | NP_727555.1 | 2 - 3 | 1769 | 72 | ||
GenBank mRNA | AY069304.1 | - | 2 - 3 | 1526 | 72 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
