Assay Details
Gene Symbol: mGluR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272147.1 | NP_001259076.1 | 3 - 4 | 1439 | 84 | |
NM_001272148.1 | NP_001259077.1 | 3 - 4 | 1439 | 84 | ||
NM_079900.3 | NP_524639.2 | 3 - 4 | 1439 | 84 | ||
GenBank mRNA | BT088899.1 | - | 3 - 4 | 1413 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
