Assay Details
| Gene Symbol: mGluR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272147.1 | NP_001259076.1 | 7 - 8 | 2443 | 120 | |
| NM_001297789.1 | NP_001284718.1 | 6 - 7 | 1899 | 120 | ||
| NM_079900.3 | NP_524639.2 | 7 - 8 | 2443 | 120 | ||
More Information
Set Membership: |
|

mGluR
Genomic Map