Assay Details
Gene Symbol: mGluR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272147.1 | NP_001259076.1 | 8 - 9 | 2750 | 97 | |
NM_001297789.1 | NP_001284718.1 | 7 - 8 | 2206 | 97 | ||
NM_079900.3 | NP_524639.2 | 8 - 9 | 2750 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
